V1138
Greiner Cryo.s™ vials
capacity 4 mL, natural cap, vial type, id field, support rim, External thread
Synonym(s):
cryo vials, cryogenic tubes, cryogenic vials, cryrogenic storage, Greiner cryogenic vials
About This Item
Recommended Products
material
colorless tube
polypropylene tube
round bottom (self-standing)
sterility
sterile (DNAse and RNAse free)
sterile
feature
vial type id field, support rim
External thread
packaging
pack of 300 ea (50 x 6 ea)
manufacturer/tradename
Greiner 127263
parameter
-196 °C min. temp.
technique(s)
cryopreservation: suitable
cap diam.
9.8 mm
capacity
4 mL
tube diam.
12.5 mm
color
natural cap
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service