Recommended Products
description
base to neck height = 86.7mm
W × H
62.4 mm × 95.8 mm
maximum volume
216 mL
volume
200 mL
application(s)
agriculture
Looking for similar products? Visit Product Comparison Guide
General description
Glass culture vessels, without caps
Autoclavable; reusable.
Autoclavable; reusable.
Compatibility
Use Magenta B-caps (Product No. B8648) as closures
recommended
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service