All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H16N2O4 · HCl
CAS Number:
Molecular Weight:
312.75
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
storage temp.
−20°C
SMILES string
C[C@@H](O)[C@H](N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1
InChI
1S/C14H16N2O4/c1-7-5-12(18)20-11-6-9(3-4-10(7)11)16-14(19)13(15)8(2)17/h3-6,8,13,17H,15H2,1-2H3,(H,16,19)/t8-,13+/m1/s1
InChI key
OIZVQDJSQAAKEU-OQPBUACISA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service