Key Documents
T4541
Supelco
Glass tubes for tube gels
I.D. × L 0.5 mm × 7.5 cm
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
I.D. × L
0.5 mm × 7.5 cm
Features and Benefits
Although the tubes listed below have different inside diameters, they all have the same 3.0 mm outside diameter. This makes the tubes less fragile and easier to handle.
Compatibility
The 7.5 cm tubes fit the mini electrophoresis unit, and the 13.5 cm tubes fit the standard-size unit.
Choose from one of the most recent versions:
Certificates of Analysis (COA)