Skip to Content
Merck
All Photos(1)

Key Documents

SRP6170

Sigma-Aldrich

IL-2 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Aldesleukin, Lymphokine, N/A, T-cell growth factor (TCGF)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

0.2-2 ng/mL

mol wt

15 kDa (monomer, glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL2(3558)

General description

IL-2 (interleukin 2) is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. It is a type I cytokine. Recombinant human IL-2 is a 15.5kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
The IL-2 gene is mapped to human chromosome 4q27.

Application

IL-2 (interleukin 2) human has been used for the stimulation of human primary naive T lymphocytes to study the effect of microRNA-146a on IL-2 activation-induced cell death.

Biochem/physiol Actions

IL-2/IL-2R (interleukin 2/IL-2 receptor) signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK (natural killer) cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes. It is associated with recurrent oral ulcer and asthma and Mycoplasma pneumoniae infection in children. Absence of IL-2 in mice causes fatal lymphoproliferative inflammatory disease (IL-2 deficiency syndrome). IL-2 signaling involves three pathways: phosphoinositol 3-kinase (PI3-K)/AKT, Ras-MAPK (mitogen activated protein kinase), and JAK-STAT (Janus kinase - signal transducers and activators of transcription) pathways.

Physical form

Lyophilized from a PBS solution.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Associations of IL-2 and IL-4 Expression and Polymorphisms With the Risks of Mycoplasma pneumoniae Infection and Asthma in Children.
Archivos de bronconeumologia, 51, 571-578 (2015)
Family based association analysis of the IL2 and IL15 genes in allergic disorders.
Christensen U, et al.
European Journal of Human Genetics, 14(2), 227-227 (2006)
An emerging player in the adaptive immune response: microRNA-146a is a modulator of IL-2 expression and activation-induced cell death in T lymphocytes.
Curtale G
Blood, 115, 265-273 (2010)
Prostaglandin E2 (PGE2)-dependent suppression of interleukin alpha (IL-2) production in patients with major trauma.
Faist E
The Journal of Trauma, 27, 837-848 (1987)
Interleukin-2 augments natural killer cell activity.
Henney CS
Nature, 291, 335-338 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service