SRP4913
Nesfatin-1
recombinant, expressed in E. coli, ≥95% (SDS-PAGE)
Synonym(s):
Nesfatin 1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
recombinant
expressed in E. coli
Assay
≥95% (SDS-PAGE)
form
lyophilized
mol wt
~8.0 kDa
packaging
pkg of 10 μg
impurities
endotoxin, tested
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Nucb2(53322)
General description
Nesfatin-1, a polypeptide produced by cleavage of the NEFA/nucleobindin-2 (NUCB2), is a secreted protein, expressed in the appetite-control hypothalamic nuclei. Nesfatin is involved in the regulation of the feeding behavior. Intracerebroventicular injection of nesfatin-1 reduces body weight in rats be affecting the animal food intake.
Physical form
Lyophilized from PBS.
Reconstitution
Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute with sterile water to a concentration of 0.1-1 mg/mL. Further dilutions should be made with medium containing 0.2% BSA.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service