SML3058
Diquafosol tetrasodium
>95% (HPLC)
Synonym(s):
Diuridine 5′-tetraphosphate, sodium salt, INS365, KPY 998, P ,P -Diuridine 5′-tetraphosphate tetrasodium salt, Up4U 4Na, Uridine 5′-(pentahydrogen tetraphosphate), P′′′→5′-ester with uridine, sodium salt (1:4), Diuridine 5′-tetraphosphate, Up4U tetrasodium
About This Item
Recommended Products
Quality Level
Assay
>95% (HPLC)
form
powder
optical activity
[α]/D -12.0 to -8.0°, c = 1.0 in H2O
color
white to beige
solubility
H2O: 2 mg/mL, clear
storage temp.
−20°C
SMILES string
[Na].[Na].[Na].[Na].[P](=O)(O[P](=O)(O[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O)N4C=CC(=O)NC4=O)O)O)(O[P](=O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)N2C=CC(=O)NC2=O)O)O
InChI
1S/C18H26N4O23P4.4Na/c23-9-1-3-21(17(29)19-9)15-13(27)11(25)7(41-15)5-39-46(31,32)43-48(35,36)45-49(37,38)44-47(33,34)40-6-8-12(26)14(28)16(42-8)22-4-2-10(24)20-18(22)30;;;;/h1-4,7-8,11-16,25-28H,5-6H2,(H,31,32)(H,33,34)(H,35,36)(H,37,38)(H,19,23,29)(H,20
InChI key
MHPMESDJCUICEY-FNXFGIETSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Diagnostic Implications
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service