Skip to Content
Merck
All Photos(1)

Key Documents

SML1835

Sigma-Aldrich

DSHN

≥98% (HPLC)

Synonym(s):

5-(Diethylsulfamoyl)-3-hydroxynaphthalene-2-carboxylic acid, 5-[(Diethylamino)sulfonyl]-3-hydroxy-2-naphthalenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17NO5S
CAS Number:
Molecular Weight:
323.36
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)S(C1=CC=CC2=CC(C(O)=O)=C(O)C=C21)(=O)=O

Biochem/physiol Actions

DSHN is a potent and selective activator of SHP (small heterodimer partner, NR0B2) that transcriptionally activates SHP mRNA and stabilized the SHP protein by preventing its ubiquitination and degradation. DSHN potently inhibits HCC cell migration and invasivnes by suppression of CCL2 expression.
DSHN is a potent and selective activator of SHP (small heterodimer partner, NR0B2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihong Yang et al.
Molecular cancer therapeutics, 15(10), 2294-2301 (2016-08-04)
Small heterodimer partner (SHP, NR0B2) is a nuclear orphan receptor without endogenous ligands. Due to its crucial inhibitory role in liver cancer, it is of importance to identify small molecule agonists of SHP. As such, we initiated a probe discovery

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service