Skip to Content
Merck
All Photos(1)

Key Documents

SML1557

Sigma-Aldrich

LP99

≥98% (HPLC)

Synonym(s):

(2R,3S)-LP99, N-(2R,3S)-2-(4-Chlorophenyl)-1-(1,4-dimethyl-2-oxo-1,2-dihydroquinolin-7-yl)-6-oxopiperidin-3-yl)-2-methylpropane-1-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30ClN3O4S
CAS Number:
Molecular Weight:
516.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C1=CC=C(N2C(CC[C@H](NS(=O)(CC(C)C)=O)[C@H]2C3=CC=C(Cl)C=C3)=O)C=C1N4C)=CC4=O

InChI

1S/C26H30ClN3O4S/c1-16(2)15-35(33,34)28-22-11-12-24(31)30(26(22)18-5-7-19(27)8-6-18)20-9-10-21-17(3)13-25(32)29(4)23(21)14-20/h5-10,13-14,16,22,26,28H,11-12,15H2,1-4H3/t22-,26+/m0/s1

InChI key

LVDRREOUMKACNJ-BKMJKUGQSA-N

Biochem/physiol Actions

LP99 is a potent and selective bromodomain BRD9 and BRD7 inhibitor with a higher potency for BRD9 (Kd = 99 nM) than for BRD7 (Kd = 909 nM), and no inhibition for any of the 48 other BRDs tested. LP99 was shown to disrupt BRD7 and BRD9 binding to chromatin in cells. LP99 also inhibited interleukin 6 (IL-6) secretion, suggesting that BRD7/9 plays a role in the regulation of pro-inflammatory cytokine secretion. LP99 is not cell cytotoxic up to 25 μM. For full characterization details, please visit the LP99 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
LP99 is a potent and selective bromodomain BRD9 and BRD7 inhibitor.

Features and Benefits

LP99 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

LP99 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the LP99 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service