Skip to Content
Merck
All Photos(1)

Key Documents

SML1386

Sigma-Aldrich

Calindol hydrochloride

≥98% (HPLC)

Synonym(s):

(R)-2-[[[1-(1-Naphthyl)ethyl]amino]methyl]-1H-indole hydrochloride, N-[(1R)-1-(1-naphthalenyl)ethyl]-1H-Indole-2-methanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20N2 · HCl
CAS Number:
Molecular Weight:
336.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[H]Cl.C[C@@H](NCC1=CC2=C(N1)C=CC=C2)C3=CC=CC4=C3C=CC=C4

InChI

1S/C21H20N2.ClH/c1-15(19-11-6-9-16-7-2-4-10-20(16)19)22-14-18-13-17-8-3-5-12-21(17)23-18;/h2-13,15,22-23H,14H2,1H3;1H/t15-;/m1./s1

InChI key

KFILKQPBQZIRST-XFULWGLBSA-N

Biochem/physiol Actions

Calcimimetic; Ca Sensing Receptor (CaSR) Agonist
Calindol is a calcimimetic Ca Sensing Receptor (CaSR) agonist. It is a positive allosteric modulator (PAM) with an EC50 value of 1 μM and a maximum stimulation of 130% of the 10mM Ca2+ activation in triggering PI hydrolysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service