Skip to Content
Merck
All Photos(1)

Key Documents

SML0478

Sigma-Aldrich

MEL-3 hydrochloride

≥98% (HPLC)

Synonym(s):

5,6-Dihydro-8-(1-methylethyl)-4H-pyrazino[3,2,1-jk]carbazole, 8-(Propan-2-yl)-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2 · HCl
CAS Number:
Molecular Weight:
286.80
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

MEL-3 is a potent androgen receptor antagonist from a new structural chemical class. MEL-3 has been shown to inhibit the proliferation of both androgen-dependent and androgen-independent AR-positive prostate cancer cell lines. MEL-3 was active against two mutant receptors (AR T877A and ARW741C) that have been found in patients who have become resistant to bicalutamide and hydroxyflutamide, two standard antiandrogens used in prostate cancer therapy.
MEL-3 is a potent androgen receptor antagonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service