Skip to Content
Merck
All Photos(1)

Documents

SML0105

Sigma-Aldrich

Cinnamyl-3,4-dihydroxy-α-cyanocinnamate

≥98% (HPLC)

Synonym(s):

CDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO4
CAS Number:
Molecular Weight:
321.33
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow

solubility

DMSO: ≥15 mg/mL

storage temp.

room temp

SMILES string

Oc1ccc(cc1O)\C=C(/C#N)C(=O)OC\C=C\c2ccccc2

InChI

1S/C19H15NO4/c20-13-16(11-15-8-9-17(21)18(22)12-15)19(23)24-10-4-7-14-5-2-1-3-6-14/h1-9,11-12,21-22H,10H2/b7-4+,16-11+

InChI key

XGHYFEJMJXGPGN-UYHGDYIZSA-N

Biochem/physiol Actions

Cinnamyl-3,4-dihydroxy-α-cyanocinnamate (CDC) is a potent and direct inhibitor of 5-LO (5-Lipoxygenase) that reduces 5-LO activity in cell-free assays. CDC also inhibits 12-LO and 15-LO

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service