Skip to Content
Merck
All Photos(1)

Key Documents

SMB00328

Sigma-Aldrich

Fmoc-L-Tyr-(β-D-Gal(Ac)4)-OH

≥95% (HPLC)

Synonym(s):

N-α-(9-Fluorenylmethyloxycarbonyl)-O-β-(2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl)-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H39NO14
Molecular Weight:
733.71
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (HPLC)

form

powder

functional group

Fmoc

storage temp.

−20°C

SMILES string

OC([C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC4=CC=C(O[C@@H]5O[C@H](COC(C)=O)[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]5OC(C)=O)C=C4)=O

InChI

1S/C38H39NO14/c1-20(40)47-19-32-33(49-21(2)41)34(50-22(3)42)35(51-23(4)43)37(53-32)52-25-15-13-24(14-16-25)17-31(36(44)45)39-38(46)48-18-30-28-11-7-5-9-26(28)27-10-6-8-12-29(27)30/h5-16,30-35,37H,17-19H2,1-4H3,(H,39,46)(H,44,45)/t31-,32?,33-,34-,35?,37+/m0/s1

InChI key

PSSMFCODGOWOEB-ROIKGVPBSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service