Skip to Content
Merck
All Photos(1)

Documents

SMB00153

Sigma-Aldrich

Asperulosidic acid

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O12
CAS Number:
Molecular Weight:
432.38
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(=O)OCC1=C[C@H](O)[C@H]2[C@@H]1[C@@H](OC=C2C(O)=O)O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O

InChI

1S/C18H24O12/c1-6(20)27-4-7-2-9(21)12-8(16(25)26)5-28-17(11(7)12)30-18-15(24)14(23)13(22)10(3-19)29-18/h2,5,9-15,17-19,21-24H,3-4H2,1H3,(H,25,26)/t9-,10+,11+,12-,13+,14-,15+,17-,18-/m0/s1

InChI key

DGDWCRWJRNMRKX-DILZHRMZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingzhen He et al.
Journal of chromatography. A, 1554, 81-91 (2018-04-25)
Efficient targeted identification of chemical constituents from traditional Chinese medicine is still a major challenge. In this study, we used a characteristic ion filtering strategy to characterize compounds of Eucommia ulmoides Oliv. by ultra-high performance liquid chromatography quadrupole time of
Heyun Zhu et al.
Journal of pharmaceutical and biomedical analysis, 111, 277-287 (2015-04-29)
Zhi-Zi-Da-Huang decoction (ZZDHD), consisting of Gardenia jasminoides Ellis, Rheum palmatum L., Citrus aurantium L. and Sojae Semen Praeparatum, is a widely used traditional Chinese medicine preparation for the treatment of acute or chronic hepatic diseases. In the present study, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service