All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C28H40N6O11
Molecular Weight:
636.65
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥75%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ac-Leu-Glu-Val-Asp-pNA
Application
Caspase 4, a cysteine protease activator of inflammatory processes, may be detected using a variety of chromogenic and fluorogenic peptide substrates built around the LEVD (leu-glu-val-asp) sequence such as the chromogenic substrate Ac-LEVD-pNa (Acetyl-Leu-Glu-Val-Asp-p-nitroanalide).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zong-Mei Bian et al.
Investigative ophthalmology & visual science, 50(12), 6006-6014 (2009-08-01)
To investigate the functional involvement of caspase-4 in human retinal pigment epithelial (hRPE) cells. Expression and activation of caspase-4 in hRPE cells were measured after stimulation with proinflammatory agents IL-1beta (2 ng/mL), TNF-alpha (20 ng/mL), lipopolysaccharide (1000 ng/mL), interferon-gamma (500
Olga Burmistrova et al.
Cancer letters, 309(1), 71-77 (2011-06-11)
Flavonoids are naturally occurring polyphenolic compounds and are among the most promising anticancer agents. Here we demonstrate that the flavonoid derivative astragalin heptaacetate (AHA) induces cell death. This was prevented by the non-specific caspase inhibitors z-VAD-fmk and Q-VD-OPH, and reduced
Akiko Yamamuro et al.
Journal of pharmacological sciences, 115(2), 239-243 (2011-02-02)
The present study investigated the function of caspase-4 in endoplasmic reticulum (ER) stress-induced apoptosis in human neuronal cell line SH-SY5Y. Tunicamycin, which is known to induce ER stress, activated both caspase-9 and caspase-4, and the activation of caspase-4 preceded that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service