Skip to Content
Merck
All Photos(2)

Documents

SAB5500122

Sigma-Aldrich

Anti-Kappa Light Chain antibody, Rabbit monoclonal

clone SP148, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP148, monoclonal

species reactivity

human (tested)

technique(s)

immunoblotting: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGKC(3514)

General description

Kappa light chains are polypeptide chains resided in the cell membrane and cytoplasmic regions of normal B cells and plasma cells. The combination of kappa light chains and heavy chains forms immunoglobulin molecules. There are two classes of light chains found in immunoglobulins, kappa light chains and lambda light chains. Individual lymphoid cell only contains a single light chain class. This clonal restriction may be used to indicate the polyclonal or monoclonal nature of B cell and plasma cell populations.The ratio of kappa to lambda is about 2:1. The level of kappa or lambda can be greatly elevated in serum of patients with multiple myeloma or other B cell malignancies.

Immunogen

Free kappa light chain from pooled human IgG lambda myeloma sera.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service