Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501885

Sigma-Aldrich

Anti-MLK3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

M3K11, MAP3K11, MAPKKK11, kinase MLK3, mitogen activated protein kinase kinase kinase 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 92 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP3K11(4296)

General description

Anti-MLK3 Antibody detects endogenous levels of total MLK3 protein.
Mitogen-activated protein kinase kinase kinase 11 (MAP3K11) or mixed lineage kinase 3 (MLK3) possesses an amino-terminal Src-homology-3 (SH3) domain followed by a kinase domain, a leucine-zipper region, a Cdc42/RAC interacting motif (CRIB) and a carboxy terminal proline/serine/threonine-rich domain. The gene encoding this serine/threonine protein kinase is localized on human chromosome 11q13.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human MLK3.

Immunogen Range: 640-689

Biochem/physiol Actions

Mitogen-activated protein kinase kinase kinase 11 (MAP3K11), also known as mixed lineage kinase 3 (MLK3), phosphorylates c-Jun N-terminal kinases. It acts as a modulator of the mitogen-activated protein kinase (MAPK) pathway. The protein is associated with various cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepatitis C virus NS5A inhibits mixed lineage kinase 3 to block apoptosis.
Amako Y, et al.
The Journal of Biological Chemistry, 288(34), 24753-24763 (2013)
Dissecting the signaling pathways associated with the oncogenic activity of MLK3 P252H mutation.
Velho S, et al.
BMC Cancer, 14(1), 182-182 (2014)
MLK3 is part of a feedback mechanism that regulates different cellular responses to reactive oxygen species.
Lee HS, et al.
Science Signaling, 7(328), ra52-ra52 (2014)
Wayne Bowden et al.
Molecular human reproduction, 15(9), 563-568 (2009-07-02)
Uterine leiomyomas (ULs) are benign monoclonal tumors originating from myometrial tissue in the uterus. Genetic pathways that lead to myometrial transformation into leiomyomas are largely unknown. Approximately 40% of ULs are karyotypically abnormal by G-banding; however, the remaining 60% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service