Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200137

Sigma-Aldrich

Anti-ALS2 (N-terminal region) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-ALS2CR6, Anti-ALSIN, Anti-ALSJ, Anti-IAHSP, Anti-PLSJ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~185 kDa

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

indirect immunofluorescence: 2-4 μg/mL using NIH3T3 cells
western blot: 1.5-3.0 μg/mL using HEK-293T cell lysate over expressing human ALS2, and rat cerebellum extract (S1 fraction).

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Als2(74018)
rat ... Als2(363235)

General description

Amyotrophic lateral sclerosis (ALS)2 gene codes for 184 kDa alsin protein. Alsin is characterized with the three putative guanine exchange factor (GEF) domains such as, RCC1-like domain (RLD), the Dbl and pleckstrin homology (DH/PH) domain, and a vacuolar protein sorting 9 (VPS9) domain. In addition, ALS2 also contains eight consecutive membrane occupation and recognition nexus (MORN) motifs. ALS2 mRNA is ubiquitously expressed in the CNS and non-neuronal tissues, with the highest expression in the cerebellum and kidney.

Specificity

Anti-ALS2 (N-terminal region) specifically recognizes human, rat, and mouse ALS2.

Application

Anti-ALS2 (N-terminal region) antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Amyotrophic lateral sclerosis (ALS)2 has been shown to mediate the activation of Rab5 and Rac1/PACK1. In the nervous system, ALS2 is preferentially associated with the cytoplasmic side of endosome membrane, it modulates endosome membrane trafficking and promotes neurite growth in neuronal cultures. Loss-of function mutations in the ALS2 gene leads to the development of recessive motor neuron diseases, including forms of ALS (ALS2), juvenile PLS (PLSJ) and hereditary spastic paraplegia (HSP).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service