Skip to Content
Merck
All Photos(1)

Key Documents

SAB2103790

Sigma-Aldrich

Anti-RNPC3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ20008, Anti-FLJ25070, Anti-KIAA183, Anti-RBM40, Anti-RNP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNPC3(55599)

Immunogen

Synthetic peptide directed towards the middle region of human RNPC3

Biochem/physiol Actions

RNPC3 is an RNA-binding protein involved in pre-mRNA splicing. RNPC3 participates in pre-mRNA U12-dependent splicing. RNPC3 binds to the 3′-stem-loop of m7G-capped U12 snRNA.Two types of spliceosomes catalyze splicing of pre-mRNAs. The major U2-type spliceosome is found in all eukaryotes and removes U2-type introns, which represent more than 99% of pre-mRNA introns. The minor U12-type spliceosome is found in some eukaryotes and removes U12-type introns, which are rare and have distinct splice consensus signals. The U12-type spliceosome consists of several small nuclear RNAs and associated proteins. This gene encodes a 65K protein that is a component of the U12-type spliceosome. This protein contains two RNA recognition motifs (RRMs), suggesting that it may contact one of the small nuclear RNAs of the minor spliceosome.

Sequence

Synthetic peptide located within the following region: LHAPLPPTSPQPPEEPPLPDEDEELSSEESEYESTDDEDRQRMNKLMELA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexis Mathian et al.
Arthritis & rheumatology (Hoboken, N.J.), 71(5), 756-765 (2018-12-07)
No simple or standardized assay is available to quantify interferon-α (IFNα) in routine clinical practice. Single-molecule array (Simoa) digital enzyme-linked immunosorbent assay (ELISA) technology enables direct IFNα quantification at attomolar (femtogram per milliliter [fg/ml]) concentrations. This study was undertaken to
Weiwei Fu et al.
The Journal of experimental medicine, 215(3), 815-825 (2018-01-31)
T follicular regulatory (Tfr) cells are a new subset of regulatory T (T reg) cells localized in the germinal center to limit the humoral response. Until now, the physiological function of Tfr cells has been largely unknown. In this study
Jae-Seung Moon et al.
Clinical rheumatology, 37(10), 2771-2779 (2017-11-10)
We investigated the rate of rheumatoid factor (RF) false positivity at diagnosis and its influence on clinical and prognostic features and rheumatoid arthritis (RA) development during the follow-up in antineutrophil cytoplasmic antibody (ANCA)-associated vasculitis (AAV) patients without RA or other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service