Skip to Content
Merck
All Photos(1)

Documents

SAB2102460

Sigma-Aldrich

Anti-TMEM16A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ10261, Anti-ORAOV2, Anti-TAOS2, Anti-Transmembrane protein 16A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

111 kDa

species reactivity

rat, guinea pig, mouse, bovine, human, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMEM16A(55107)

Immunogen

The immunogen for anti-TMEM16A antibody: synthetic peptide derected towards the middle region of human TMEM16A

Sequence

Synthetic peptide located within the following region: HGFVNHTLSSFNVSDFQNGTAPNDPLDLGYEVQICRYKDYREPPWSENKY

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael Courjaret et al.
Journal of cell science, 129(13), 2548-2558 (2016-05-14)
Ca(2+)-activated Cl(-) channels (CaCCs) play important physiological functions in epithelia and other tissues. In frog oocytes the CaCC Ano1 regulates resting membrane potential and the block to polyspermy. Here, we show that Ano1 expression increases the oocyte surface, revealing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service