Skip to Content
Merck
All Photos(1)

Documents

SAB2100669

Sigma-Aldrich

Anti-EIF4G3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-EIF4GII, Anti-Eukaryotic translation initiation factor 4 γ, 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

174 kDa

species reactivity

bovine, rabbit, mouse, guinea pig, rat, human, dog, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EIF4G3(8672)

Immunogen

Synthetic peptide directed towards the middle region of human EIF4G3

Biochem/physiol Actions

EIF4G3 is probable component of the protein complex eIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5′-terminal secondary structure and recruitment of mRNA to the ribosome. It is thought to be a functional homolog of EIF4G1.

Sequence

Synthetic peptide located within the following region: MRGGSSKDLLDNQSQEEQRREMLETVKQLTGGVDVERNSTEAERNKTRES

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hashimoto et al.
Translational psychiatry, 4, e472-e472 (2014-10-22)
The superior frontal gyrus (SFG), an area of the brain frequently found to have reduced gray matter in patients with schizophrenia, is involved in self-awareness and emotion, which are impaired in schizophrenia. However, no genome-wide association studies of SFG volume
Sanjay Ghosh et al.
PloS one, 10(4), e0122519-e0122519 (2015-04-08)
In eukaryotes, post-transcriptional regulation of gene expression has a key role in many cellular and developmental processes. Spermatogenesis involves a complex developmental program that includes changes in cell cycle dynamics and dramatic cellular remodeling. Translational control is critical for spermatogenesis
A Gradi et al.
Molecular and cellular biology, 18(1), 334-342 (1998-01-07)
Mammalian eukaryotic translation initiation factor 4F (eIF4F) is a cap-binding protein complex consisting of three subunits: eIF4E, eIF4A, and eIF4G. In yeast and plants, two related eIF4G species are encoded by two different genes. To date, however, only one functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service