Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410918

Sigma-Aldrich

Anti-PDCD2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC12347, RP8, ZMYND7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 38.6 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDCD2(5134)

General description

This gene encodes a nuclear protein expressed in a variety of tissues. The rat homolog, Rp8, is transiently expressed in immature thymocytes and is thought to be involved in programmed cell death. Expression of the human gene has been shown to be repressed by BCL6, a transcriptional repressor required for lymph node germinal center development, suggesting that BCL6 regulates apoptosis by its effects on PDCD2. This gene is closely linked on chromosome 6 to the gene for TBP, the TATA binding protein. Two transcripts encoding different proteins have been identified. (provided by RefSeq)

Immunogen

PDCD2 (NP_002589.2, 1 a.a. ~ 344 a.a) full-length human protein.

Sequence
MAAAGARPVELGFAESAPAWRLRSEQFPSKVGGRPAWLGAAGLPGPQALACELCGRPLSFLLQVYAPLPGRPDAFHRCIFLFCCREQPCCAGLRVFRNQLPRKNDFYSYEPPSENPPPETGESVCLQLKSGAHLCRVCGCLGPKTCSRCHKAYYCSKEHQTLDWRLGHKQACAQPDHLDHIIPDHNFLFPEFEIVIETEDEIMPEVVEKEDYSEIIGSMGEALEEELDSMAKHESREDKIFQKFKTQIALEPEQILRYGRGIAPIWISGENIPQEKDIPDCPCGAKRILEFQVMPQLLNYLKADRLGKSIDWGILAVFTCAESCSLGTGYTEEFVWKQDVTDTP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service