Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306674

Sigma-Aldrich

ANTI-ALDOLASE (ALDOA)(N-TERMINAL) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ALDA, ALDOA, Fructose-bisphosphate aldolase A, Lung cancer antigen NY-LU-1, Muscle-type aldolase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
IF
IHC
WB
species reactivity:
human
technique(s):
immunofluorescence: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000
citations:
4
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39420 Da

species reactivity

human

technique(s)

immunofluorescence: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDOA(226)

General description

Aldolase A, fructose-bisphosphate (ALDOA) is a ubiquitous glycolytic enzyme expressed in developing embryo and in adult muscle. It is involved in a wide range of cellular functions such as maintenance of striated muscle contraction, cell shape and mobility, actin filament organization and ATP biosynthetic process.

Application

ANTI-ALDOLASE (ALDOA)(N-TERMINAL) antibody produced in rabbit is suitable for immunofluorescence (1:10-1:50), immunohistochemistry (1:10-1:50), and western blot (1:1000) applications.

Biochem/physiol Actions

In glycolysis, ALDOA catalyzes the reversible reaction of fructose-1,6-bisphosphate to glyceraldehydes-3-phosphate and dihydroxyacetone phosphate. High expression level of ALDOA is reported in various forms of malignant cancers, including human lung squamous, renal cell and hepatocellular carcinomas. ALDOA deficiency causes myopathy and hemolytic anemia.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Miwa et al.
American journal of hematology, 11(4), 425-437 (1981-12-01)
Two cases of red cell aldolase deficiency associated with congenital nonspherocytic hemolytic anemia are reported. The proband is a fourteen-month-old Japanese boy. Consanguineous marriage was not proven but probable in this family, as the parents were born in the same
Tamami Morisaki et al.
PloS one, 9(11), e110736-e110736 (2014-11-08)
Cancer stem cells (CSCs) are responsible for cancer progression, metastasis, and recurrence. To date, the specific markers of CSCs remain undiscovered. The aim of this study was to identify novel biomarkers of gastric CSCs for clinical diagnosis using proteomics technology.
H Kishi et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(23), 8623-8627 (1987-12-01)
Fructose-1,6-bisphosphate aldolase A (fructose-bisphosphate aldolase; EC 4.1.2.13) deficiency is an autosomal recessive disorder associated with hereditary hemolytic anemia. To clarify the molecular mechanism of the deficiency at the nucleotide level, we have cloned aldolase A cDNA from a patient's poly(A)+
Sha Du et al.
PloS one, 9(1), e85804-e85804 (2014-01-28)
Fructose-bisphosphate aldolase A (ALDOA) is a key enzyme in glycolysis and is responsible for catalyzing the reversible conversion of fructose-1,6-bisphosphate to glyceraldehydes-3-phosphate and dihydroxyacetone phosphate. ALDOA contributes to various cellular functions such as muscle maintenance, regulation of cell shape and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service