Skip to Content
Merck
All Photos(1)

Documents

S5944

Sigma-Aldrich

S-15176 difumarate salt

≥98% (HPLC), solid

Synonym(s):

N-[(3,5-Di-tert-butyl-4-hydroxy-1-thiophenyl)]-3-propyl-N′-(2,3,4-trimethoxybenzyl)piperazine difumarate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C31H48N2O4S · 2 C4H4O4
CAS Number:
Molecular Weight:
776.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 16 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.COc1ccc(CN2CCN(CCCSc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)CC2)c(OC)c1OC

InChI

1S/C31H48N2O4S.2C4H4O4/c1-30(2,3)24-19-23(20-25(27(24)34)31(4,5)6)38-18-10-13-32-14-16-33(17-15-32)21-22-11-12-26(35-7)29(37-9)28(22)36-8;2*5-3(6)1-2-4(7)8/h11-12,19-20,34H,10,13-18,21H2,1-9H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1+

InChI key

ANXPOPNCJIJXJW-LVEZLNDCSA-N

Application

The protonophoric activity of S-15176 difumarate salt was studied in mitochondria isolated from livers of Wistar rats.5

Biochem/physiol Actions

Antioxidant and anti-ischemic agent. Also inhibits mitochondrial permeability transition, prevents the early step in apoptosis by preventing collapse of the electrochemical gradient across the mitochondrial membrane. IC50 for in vitro lipid peroxidation is 0.3 μM. IC50 for carnitine palmitoyltransferase (CPT-1) in heart homogenate is 16.8 μM. The shift from fatty acid to glucose oxidation may contribute to anti-ischemic effect.
S-15176 suppresses the mitochondrial permeability transition in response to calcium ions and inorganic phosphate and inhibits the release of cytochrome c in response to silver ions. It exhibits weak protonophoric activity.5

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aziz Elimadi et al.
European journal of pharmacology, 468(2), 93-101 (2003-05-14)
Mitochondrial Ca(2+) accumulation can induce a sudden increase in the permeability of the inner membrane. This phenomenon is due to the generation of a large nonselective ion channel, termed the permeability transition pore (PTP), which contributes to cellular injury during
A Settaf et al.
European journal of pharmacology, 406(2), 281-292 (2000-10-06)
The protective effect of N-[(3, 5-di-tertiobutyl-4-hydroxy-1-thiophenyl)]-3-propyl-N'-(2,3, 4-trimethoxybenzyl)piperazine (S-15176) on liver injury induced by warm ischemia-reperfusion was investigated using a rat model. Animals were subjected to 2 h of ischemia followed by different reperfusion times. Hepatocyte integrity was assessed by measuring
M Hamdan et al.
Pharmacological research, 44(2), 99-104 (2001-08-23)
The purpose of this study was to investigate the possible effect of the trimetazidine derivative S-15176 on carnitine palmitoyltransferase1 (CPT-1) activity in rat heart and liver mitochondria. S-15176 was compared with the other antianginal agents amiodarone, perhexiline and trimetazidine, which
Satoshi Kawashima et al.
Molecular and cellular biochemistry, 358(1-2), 45-51 (2011-06-21)
A recent report has described that S-15176 (N-[(3,5-di-tert-butyl-4-hydroxy-1-thiophenyl)]-3-propyl-N'-(2,3,4-trimethoxybenzyl) piperazine), an anti-ischemic agent, inhibits the mitochondrial permeability transition (PT) induced by not only Ca(2+) and inorganic phosphate, but also by tert-butylhydroperoxide or phenylarsine oxide [Morin et al. (Biochem Pharmacol 72:911-918, 2006)].
Heinz Rupp et al.
Herz, 27(7), 621-636 (2002-11-20)
Partial fatty acid oxidation inhibitors have raised great interest since they are expected to counteract a dysregulated gene expression of hypertrophied cardiocytes. Some of these compounds have been developed for treating non-insulin-dependent diabetes mellitus and stable angina pectoris. A shift

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service