Skip to Content
Merck
All Photos(1)

Key Documents

RAB1078

Sigma-Aldrich

Human PYY ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.15 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PYY(5697)

General description

This ELISA antibody pair detects human PYY. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asra Iftikhar et al.
Oxidative medicine and cellular longevity, 2020, 9020219-9020219 (2020-04-08)
Caesalpinia bonduc has been used in herbal medicines for the treatment of a wide range of diseases from decades. The present study has explored the remedial potential and underlying mechanism of polyphenol extract of Caesalpinia bonduc in alloxanized diabetic rats.
Steven K Malin et al.
Physiology & behavior, 223, 112978-112978 (2020-06-01)
Caloric restriction is suggested to increase hunger, in part, through complex interactions of hormones and behavior that contribute to challenges in long-term weight loss. Although intense exercise may attenuate appetite, no data exist testing the effects of interval exercise (INT)
Charmaine D Espinosa et al.
Journal of animal science, 98(10) (2020-09-13)
Two experiments were conducted to test the hypothesis that torula yeast may replace animal and plant proteins in diets for weanling pigs without negatively impacting growth performance or blood characteristics. In exp. 1, 128 weanling pigs (6.71 ± 0.76 kg)
Ipek Acar et al.
Nutritional neuroscience, 23(6), 481-489 (2018-09-18)
Objectives: The calcium-sensing receptor (CaSR), the major sensor of extracellular Ca2+, is expressed in various tissues, including the gastrointestinal tract. Although the essential ligand of CaSR is calcium, its activity can be regulated by aromatic L-amino acids. The expression of
Min Tian et al.
Journal of agricultural and food chemistry, 67(24), 6809-6818 (2019-05-29)
Strategies to increase feed intake are of great importance for producing more meat in swine production. Intestinal and hypothalamic amino acid receptors are found to largely participate in feed intake regulation. The purpose of the current research is to study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service