Skip to Content
Merck
All Photos(1)

Documents

RA44450-EP

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

A-4-44, aerosol-tight, with 4 buckets for 2×50 mL conical tubes

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

to hold 0.2 mL to 100 mL tubes
aerosol-tight
high-speed swing-bucket rotor
with 4 buckets for 2×50 mL conical tubes

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022637461 (NA)
Eppendorf® 5804719000 (EU)

parameter

4,500 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

  • A high-speed swing-bucket rotor (A-4-44) holds 0.2 mL to 100 mL tubes.
  • Rotors, buckets, lid and adapters are autoclavable at 121 °C, 20 min.
  • Optional aerosol-tight caps are available for the safe centrifugation of hazardous samples.
  • Rotor lifetime: max. 34,000 cycles.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service