Skip to Content
Merck
All Photos(1)

Key Documents

R115

Sigma-Aldrich

Reactive Blue 2

Synonym(s):

1-Amino-4-[[4-[[4-chloro-6-[[3 (or 4)-sulfophenyl]amino]-1,3,5-triazin-2-yl]amino]-3-sulfophenyl]amino]-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid, Basilen Blue E-3G, Cibacron Blue F3G-A, Cibacron Blue F3GA, GST Inhibitor-1, Procion® Blue H-B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H17ClN7Na3O11S3
CAS Number:
Molecular Weight:
840.10
Colour Index Number:
61211
Beilstein:
6561550
EC Number:
UNSPSC Code:
12171500
PubChem Substance ID:

color

dark blue

solubility

H2O: soluble

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].Nc1c(cc(Nc2ccc(Nc3nc(Cl)nc(Nc4ccc(cc4)S([O-])(=O)=O)n3)c(c2)S([O-])(=O)=O)c5C(=O)c6ccccc6C(=O)c15)S([O-])(=O)=O.Nc7c(cc(Nc8ccc(Nc9nc(Cl)nc(Nc%10cccc(c%10)S([O-])(=O)=O)n9)c(c8)S([O-])(=O)=O)c%11C(=O)c%12ccccc%12C(=O)c7%11)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

P2Y purinoceptor antagonist; most potent antagonist for ATP-activated channels.

Legal Information

Procion is a registered trademark of Dystar Textilfarben GmbH & Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Anikina et al.
Bulletin of experimental biology and medicine, 152(6), 672-674 (2012-07-18)
We studied the effect of uridine 5'-triphosphate in concentrations of 10(-10)-10(-6) M on myocardial contractile activity in 7-100-day-old rats. Analysis of isometric contraction of myocardial strips showed that uridine 5'-triphosphate reduced the strength of myocardial contraction in rats of all
Mehmet Odabası
Preparative biochemistry & biotechnology, 41(3), 287-304 (2011-06-11)
Cibacron Blue F3GA was covalently attached onto magnetic poly(vinyl alcohol) (mPVAL) beads (100-150 μm in diameter) for human serum albumin (HSA) adsorption from human plasma. Despite low nonspecific adsorption of HSA on mPVAL beads, Cibacron Blue F3GA attachment significantly increased
K Inoue et al.
British journal of pharmacology, 102(4), 851-854 (1991-04-01)
1. The effects of reactive blue 2 and brilliant blue G, which have been shown to block extracellular ATP-evoked responses, were investigated to discover whether these compounds act as P2-purinoceptor antagonists in PC12 phaeochromocytoma cells. 2. Reactive blue 2 (10
Angel Luis García-Villalón et al.
European journal of pharmacology, 660(2-3), 394-401 (2011-04-26)
Diadenosine tetraphosphate (AP4A) is a vasoactive mediator that may be released from platelet granules and that may reach higher plasma concentrations during coronary ischemia-reperfusion. The objective of this study was to analyze its coronary effects in such conditions. To this
Miao Li et al.
Protein and peptide letters, 20(4), 459-466 (2012-09-29)
From the fresh fruiting bodies of the mushroom Pleurotus eryngii a phytase with a molecular mass of 14 kDa was isolated. The isolation protocol entailed ion exchange chromatography on DEAE-cellulose and CM-cellulose, affinity chromatography on Affi-gel blue gel, and ion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service