R104
Rauwolscine hydrochloride
powder
Synonym(s):
α-Yohimbine hydrochloride, 17α-Hydroxy-20α-yohimban-16 β-carboxylic acid methyl ester hydrochloride
About This Item
form
powder
optical activity
[α]25/D +52.13°, c = 1 in H2O
color
white to off-white
solubility
H2O: soluble
SMILES string
Cl.COC(=O)[C@@H]1[C@@H](O)CCC2CN3CCc4c([nH]c5ccccc45)C3CC12
Gene Information
human ... ADRA2A(150) , ADRA2B(151) , ADRA2C(152) , HTR1A(3350)
Biochem/physiol Actions
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service