Skip to Content
Merck
All Photos(1)

Key Documents

P8962

Sigma-Aldrich

Phenanthrene-9-14C

methanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

form

methanol solution

extent of labeling

5-15 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)c[14cH]c3ccccc23

Looking for similar products? Visit Product Comparison Guide

Packaging

sealed ampule

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service