Skip to Content
Merck
All Photos(1)

Documents

P8893

Sigma-Aldrich

Phorbol

≥98% (TLC), powder

Synonym(s):

4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O6
CAS Number:
Molecular Weight:
364.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

C[C@@H]1[C@@H](O)[C@]2(O)[C@H]([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mammary tumor promoter in experimental models.

Warning

Possibly carcinogenic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Promotion of mammary carcinogenesis and leukemogenic action by phorbol in virgin female Wistar rats.
V Armuth et al.
Cancer research, 34(10), 2704-2707 (1974-10-01)
Renoud J Marijnissen et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(4), 886-895 (2014-04-24)
The cytokine interleukin-21 (IL-21) can have both proinflammatory and immunosuppressive effects. The purpose of this study was to investigate the potential dual role of IL-21 in experimental arthritis in relation to Th17 cells. Antigen-induced arthritis (AIA) and chronic streptococcal cell
Eveline M Delemarre et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(2), 350-356 (2014-02-08)
Autologous stem cell transplantation (ASCT) induces long-term drug-free disease remission in patients with juvenile idiopathic arthritis. This study was undertaken to further unravel the immunologic mechanisms underlying ASCT by using a mouse model of proteoglycan-induced arthritis (PGIA). For initiation of
Takehiro Takahashi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(1), 254-265 (2014-10-11)
Bleomycin-induced fibrosis and the tight skin (TSK/+) mouse are well-established experimental murine models of human systemic sclerosis (SSc). Growing evidence has demonstrated the pivotal role of Toll-like receptors (TLRs) in several autoimmune inflammatory diseases, including SSc. This study was undertaken
Frederique M Moret et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(5), 1176-1184 (2014-05-02)
To determine the levels of thymic stromal lymphopoietin (TSLP) and the numbers of TSLP receptor (TSLPR)-expressing CD1c+ (blood dendritic cell antigen 1-positive) myeloid dendritic cells (MDCs) in the joints as compared with the peripheral blood (PB) of patients with rheumatoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service