Skip to Content
Merck
All Photos(1)

Key Documents

P4277

Supelco

(R)-(+)-1-Phenylethanol

Synonym(s):

(+)-Methyl phenyl carbinol, (R)-(+)-α-Methylbenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Beilstein:
2039798
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

bp

88-89 °C/10 mmHg (lit.)

mp

9-11 °C (lit.)

density

1.012 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](O)c1ccccc1

InChI

1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/t7-/m1/s1

InChI key

WAPNOHKVXSQRPX-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachiko Sugimoto et al.
Phytochemistry, 108, 189-195 (2014-12-03)
Three aromatic glycosides (1-3), two sulfur and nitrogen-containing compound glucosides (4, 5), and one flavonoid glycoside (6) were isolated from the leaves of Ixora undulata. Their structures were established by extensive 1D, 2D NMR, and HRESIMS experiments, and structure 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service