Skip to Content
Merck
All Photos(1)

Key Documents

P2053

Sigma-Aldrich

pGlu-Glu-Pro Amide

≥97% (HPLC)

Synonym(s):

Fertilization Promoting Peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N4O6
CAS Number:
Molecular Weight:
354.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

NC(=O)C1CCCN1C(=O)C(CCC(O)=O)NC(=O)C2CCC(=O)N2

InChI

1S/C15H22N4O6/c16-13(23)10-2-1-7-19(10)15(25)9(4-6-12(21)22)18-14(24)8-3-5-11(20)17-8/h8-10H,1-7H2,(H2,16,23)(H,17,20)(H,18,24)(H,21,22)

InChI key

HYZBGWLLSXSYLX-UHFFFAOYSA-N

Amino Acid Sequence

Glp-Glu-Pro-NH2

Biochem/physiol Actions

TRH analog that is produced in the thyroid, mammary gland and prostate. Promotes fertilization by enhancing sperm function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Ruiz-Alcaraz et al.
The Journal of clinical endocrinology and metabolism, 88(12), 6002-6007 (2003-12-13)
The TRH-like peptides pGlu-Glu-Pro amide, pGlu-Phe-Pro amide, and pGlu-Gln-Pro amide were isolated and identified some years ago, and these peptides have been proven to be present in many tissues and fluids. The presence of TRH-like immunoreactivity distinct from TRH in
Yongxin Ma et al.
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics, 20(2), 151-155 (2003-04-04)
Both fertilization promoting peptide and adenosine stimulate capacitation but inhibit spontaneous acrosome loss by modulation of the adenylyl cyclase (AC)/cAMP signal transduction pathway. This is a review aimed at analyzing the function of fertilization promoting peptide during this process. The
Molecular mechanisms during sperm capacitation.
Ian A Brewis et al.
Human fertility (Cambridge, England), 8(4), 253-261 (2006-01-06)
L R Fraser
Frontiers in bioscience : a journal and virtual library, 3, D1187-D1191 (1998-12-04)
Fertilization promoting peptide (FPP), structurally similar to thyrotrophin releasing hormone, is produced by the prostate gland and secreted into seminal plasma. Recent in vitro studies have provided evidence that FPP elicits biologically relevant responses in mouse, human and boar spermatozoa.
C K Park et al.
Animal reproduction science, 72(1-2), 83-94 (2002-07-11)
This study has evaluated the effect of fertilization-promoting peptide (FPP) on the fertilizing ability and glycosidase activity in vitro of frozen-thawed boar spermatozoa. Use of chlortetracycline (CTC) fluorescence analysis, as well as various glycosidase analyses and the oocyte penetration test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service