Skip to Content
Merck
All Photos(1)

Key Documents

P0105

Sigma-Aldrich

PM-20

≥96% (HPLC), solid

Synonym(s):

1-[1,1′-Biphenyl]-4-yl-3,4-bis[(2-hydroxyethyl)thio]-1H-Pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO4S2
CAS Number:
Molecular Weight:
401.50
MDL number:
UNSPSC Code:
12352200

Assay

≥96% (HPLC)

form

solid

color

yellow

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

OCCSC1=C(SCCO)C(=O)N(C1=O)c2ccc(cc2)-c3ccccc3

InChI

1S/C20H19NO4S2/c22-10-12-26-17-18(27-13-11-23)20(25)21(19(17)24)16-8-6-15(7-9-16)14-4-2-1-3-5-14/h1-9,22-23H,10-13H2

InChI key

RFZCIUONZTUZER-UHFFFAOYSA-N

Biochem/physiol Actions

PM-20 is a Cdc25A phosphatase inhibitor.

Caution

air sensitive

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siddhartha Kar et al.
Molecular cancer therapeutics, 5(6), 1511-1519 (2006-07-05)
We have synthesized several new phenyl maleimide compounds, which are potent growth inhibitors of several human tumor cell lines. Among these, PM-20 was the most potent with an IC50 of 700 nmol/L for Hep3B human hepatoma cell growth. Two other
Irina V Chikalovets et al.
Fish & shellfish immunology, 42(2), 503-507 (2014-12-09)
Lectins (carbohydrate-binding proteins) are well known to actively participate in the defense functions of vertebrates and invertebrates where they play an important role in the recognition of foreign particles. In this study, we investigated of in vitro antifungal activity of
Joon-Khim Loh et al.
BMC cancer, 10, 268-268 (2010-06-10)
High-grade gliomas have poor prognosis, requiring aggressive treatment. The aim of this study is to explore mitotic and centrosomal dysregulation in gliomas, which may provide novel targets for treatment. A case-control study was performed using 34 resected gliomas, which were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service