N6627
D-Norleucine
Synonym(s):
(R)-2-Aminocaproic acid, (R)-2-Aminohexanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721749
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Assay
≥98%
form
powder or crystals
technique(s)
ligand binding assay: suitable
color
white to off-white
mp
300 °C
application(s)
peptide synthesis
SMILES string
CCCC[C@@H](N)C(O)=O
InChI
1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m1/s1
InChI key
LRQKBLKVPFOOQJ-RXMQYKEDSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Norleucine is a synthetic amino acid commonly used as an internal standard.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Imbrogno et al.
General and comparative endocrinology, 240, 1-9 (2016-10-23)
Chromogranin A (CgA) is an acidic protein co-stored with catecholamines, hormones and neuropeptides in the secretory granules of endocrine, neuronal and other cell types (including cardiomyocytes). Proteolytic cleavage in the C terminus of CgA generates a 2.9kDa peptide named serpinin
Christopher T Yarnes et al.
Rapid communications in mass spectrometry : RCM, 31(8), 693-704 (2017-02-09)
Within the last decade, applications of compound-specific stable isotope analysis of nitrogen (δ We examined the relative effect of three normalization procedures, (1) internal reference calibration, (2) compound-specific calibration, and (3) scale-normalization, on the accuracy and precision of δ The
Tianliang He et al.
Oncogene (2018-06-22)
The antiviral metabolites from bacterial stress response to bacteriophage infection can maintain homeostasis of host cells, while metabolism disorder is a remarkable characteristic of tumorigenesis. In the aspect of metabolic homeostasis, therefore, the antiviral homeostasis-maintaining metabolites of bacteria may possess
Stephanie N Dupuis et al.
Journal of natural products, 74(11), 2420-2424 (2011-11-05)
Streptomyces venezuelae ISP5230 is recognized for the production of chloramphenicol and the jadomycin family of natural products. The jadomycins are angucycline natural products containing a unique oxazolone ring incorporating an amino acid present in the minimal culture media. Substitution of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service