N5378
N-(2-Nitrophenylsulfenyl)glycine (dicyclohexylammonium) salt
powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8N2O4S · C12H23N
CAS Number:
Molecular Weight:
409.54
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
form
powder
color
yellow
storage temp.
−20°C
SMILES string
C1CCC(CC1)NC2CCCCC2.OC(=O)CNSc3ccccc3[N+]([O-])=O
InChI
1S/C12H23N.C8H8N2O4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;11-8(12)5-9-15-7-4-2-1-3-6(7)10(13)14/h11-13H,1-10H2;1-4,9H,5H2,(H,11,12)
InChI key
QHKBANTYLBSWKY-UHFFFAOYSA-N
Biochem/physiol Actions
N-(2-Nitrophenylsulfenyl)glycine (dicyclohexylammonium) salt is the stable NPS derivative of glycine.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Efficient Procedure for the Preparation of Oligomer-Free N α -Fmoc Amino Acids
Nowshuddina, S., Raoa, M.N.A., and Ram Reddya, A.
Synthetic Communications, 39, 2022-2031 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service