Skip to Content
Merck
All Photos(1)

Documents

MAK449

Sigma-Aldrich

Free Amino Nitrogen Assay Kit

Sufficient for 100 colorimetric tests

Synonym(s):

FAN Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

storage condition

(No metal containers. Tightly closed.)

input

food(s)
beverage(s)

application(s)

cosmetics
food and beverages

detection method

colorimetric

storage temp.

2-8°C

Related Categories

General description

Free Amino Nitrogen (FAN) is the main source of nitrogen necessary for yeast growth and proper fermentation. Fermentation of beer and wine is processed by yeast, which synthesize proteins using available amino acids. When making beer and wine, free amino nitrogen is extracted from amino acids during the formation of the wort or must.

Application

The Free Amino Nitrogen Assay Kit may be used for:
  • Alcohol Fermentation Analysis
  • Food and Beverage Analysis
  • Yeast Proliferation Analysis

Features and Benefits

  • Efficient and Precise: Utilizes just 5 μL of sample while detecting a wide range of 0.2 to 10 mM free amino nitrogen, providing sensitivity and accuracy.
  • Streamlined Procedure: Experience a straightforward process with the addition of a single working reagent and a quick 10-minute assay reaction, minimizing time and effort required.
  • Compatibility with High-Throughput Systems: Easily integrate our kit into high-throughput handling systems, guaranteeing efficiency and precision in your laboratory operations.

Suitability

The kit is suitable for Free Amino Nitrogen determination in foods and beverages (e.g. beer, wort, wine, must, etc.).

Principle

The Free Amino Nitrogen Assay Kit measures alpha amino acids, ammonia, and end group amino nitrogens. The ninhydrin based reaction is a superior method for determining only alpha amino acids and ammonia compared to the traditional Kjeldahl, which measures nitrogen from all sources. The stable ninhydrin reagent provides a simple and accurate method for determining Free Amino Nitrogen concentrations.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service