Skip to Content
Merck
All Photos(1)

Key Documents

M0900

Sigma-Aldrich

Methyl 3-O-β-D-galactopyranosyl-β-D-galactopyranoside

~99%

Synonym(s):

β-D-Gal-[1→3]-β-D-Gal-1→OMe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O11
CAS Number:
Molecular Weight:
356.32
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

~99%

storage temp.

−20°C

SMILES string

CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@H]1O

Application

A model compound for the study of galactopyranans by NMR spectroscopy. Useful for specificity studies of antigalactopyranan monoclonal antibodies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gorin, P.A.J.,
Carbohydrate Research, 101, 13-13 (1982)
An efficient, unambiguous synthesis of methyl 3-O-beta-D-galactopyranosyl-beta-D-galactopyranoside. Further studies on the specificity of antigalactopyranan monoclonal antibodies.
P Kovác et al.
Carbohydrate research, 142(1), 158-164 (1985-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service