Skip to Content
Merck
All Photos(1)

Key Documents

L7538

Sigma-Aldrich

D-Luciferin potassium salt

Synonym(s):

4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7KN2O3S2
Molecular Weight:
318.41
MDL number:
UNSPSC Code:
12171500

mol wt

318.41

solubility

H2O: soluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

[K+].Oc1ccc2nc(sc2c1)C3=N[C@H](CS3)C([O-])=O

Biochem/physiol Actions

Water soluble substrate for firefly luciferase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service