L7271
Leukotriene C4 methyl ester
~50 μg/mL in methanol: water (7:3) containing ammonium acetate buffer (≤0.1%), pH 5.4, ~97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Assay
~97%
concentration
~50 μg/mL in methanol: water (7:3) containing ammonium acetate buffer (≤0.1%), pH 5.4
shipped in
dry ice
storage temp.
−20°C
Biochem/physiol Actions
Methyl ester form of Leukotriene C4 which induces bronchoconstriction and renal vasoconstriction.
Packaging
Packaged under Argon.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S V Galushko et al.
Bioorganicheskaia khimiia, 15(7), 952-959 (1989-07-01)
The chromatographic (RP HPLC) behaviour of leukotriene C4, its methyl ester, leukotriene A4 methyl ester and some chemicals involved in their synthesis have been investigated. Optimal conditions of separation were determined for the gradient and isocratic HPLC. Parameters of the
R Agarwal et al.
Biochemical pharmacology, 44(10), 2047-2053 (1992-11-17)
The glutathione S-transferase (GST)-dependent conjugation of reduced glutathione (GSH) with leukotriene A4 (LTA4)-methyl ester in rodent and human skin was investigated. Incubation of [3H]LTA4-methyl ester (1 nmole, approximately 200,000 dpm) with cytosol prepared from rat, mouse and human skin or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service