Skip to Content
Merck
All Photos(3)

Documents

L6037

Sigma-Aldrich

Escort II Transfection Reagent

Lipid formulation for nucleic acid transfections

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

for molecular biology

form

dried film

usage

1.5 mL sufficient for 150 transfections

storage temp.

2-8°C

General description

Escort II is a unique formulation of a proprietary cationic lipid and the neutral lipid DOPE. This liposome-forming compound is used for transfection of nucleic acids into eukaryotic cells.

Application

Suitable for transient and stable transfection of nucleic acids into cultured eukaryotic cells. Use approximately 25 μl Escort II and 5 μg DNA (in provided unique DNA Dilution buffer) per 6 cm cell culture plate. Protocol optimization provides very efficient transfection. The following cells have been successfully transfected using Escort II:

B16-F0
BHK-21
CHO-K1
COS-1
COS-7
CV-1
D. mel (S2)
F9
HepG2
HEK293
HeLa
HUVEC
K562
L929
MDCK
NIH3T3
P19
PC-12
Vero

Features and Benefits

  • Suitable for stable and transient transfection
  • High efficiency for a wide variety of cell types
  • Compatible with both serum and serum-free transfection protocols
  • Low toxicity

Components

Escort II Transfection Kit (0.75 mL) contains:
0.75 mL Escort II Reagent (L6037)
0.8 mL Hydration Buffer (B1180)
4 mL DNA Dilutent (D6813)

Escort II Transfection Kit (1.5 mL) contains:
1.5 mL Escort II Reagent (L6037)
1.6 mL Hydration Buffer (B1180)
4 mL DNA Dilutent (D6813)

Note the formulation of Escort II is confidential.

Caution

Do not freeze.

Principle

A stable complex is formed when Escort II is mixed with DNA in the absence of serum. The complexes are stable and can be directly added to the cell culture medium, where they fuse with the cell membrane, releasing the DNA into the cytoplasm. Note: complex formation is inhibited by serum, but once stable complexes have formed, the presence of serum is without consequence.

Legal Information

Escort is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service