Skip to Content
Merck
All Photos(1)

Key Documents

L5152

Sigma-Aldrich

Lewis-X trisaccharide

≥99% (HPLC)

Synonym(s):

α-L-Fuc-(1→3)-[β-D-Gal-(1→4)]-D-GlcNAc, Lex Trisaccharide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35NO15
CAS Number:
Molecular Weight:
529.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (HPLC)

storage temp.

2-8°C

SMILES string

CC1OC(OC(C(NC(C)=O)C=O)C(OC2OC(CO)C(O)C(O)C2O)C(O)CO)C(O)C(O)C1O

InChI

1S/C20H35NO15/c1-6-11(27)13(29)15(31)19(33-6)35-17(8(3-22)21-7(2)25)18(9(26)4-23)36-20-16(32)14(30)12(28)10(5-24)34-20/h3,6,8-20,23-24,26-32H,4-5H2,1-2H3,(H,21,25)

InChI key

ZAFUNKXZZPSTLA-UHFFFAOYSA-N

Biochem/physiol Actions

Lewis-X is a developmentally regulated antigen and has been suggested to be a tumor marker. At high concentrations, it inhibits P-selectin−mediated adhesion.

Other Notes

Lewis-X is the trisaccharide part of the sialyl-Lewis-X tetrasaccharide, the suggested ligand for P- and E-selectins.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilkins, W.M., et al.
ACS Symp. Ser., 519, 34-34 (1993)
Marko Haramija et al.
Rapid communications in mass spectrometry : RCM, 31(20), 1709-1719 (2017-08-15)
Affinity mass spectrometry (AMS) is an emerging tool in the field of the study of protein•carbohydrate complexes. However, experimental obstacles and data analysis are preventing faster integration of AMS methods into the glycoscience field. Here we show how analysis of
L A Lasky
Science (New York, N.Y.), 258(5084), 964-969 (1992-11-06)
Although a bewildering array of cell surface carbohydrate structures have been described, the physiological relevance of any of these complex molecules has often eluded biologists. A family of cell surface glycoproteins, the "selectins," has a characteristic ability to use some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service