Skip to Content
Merck
All Photos(1)

Key Documents

K1640

Sigma-Aldrich

Anti-Kallikrein 15 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-KLK15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~28 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... KLK15(55554)
mouse ... Klk15(317652)
rat ... Klk15(292873)

Immunogen

synthetic peptide corresponding to residues 75-87 of human Kallikrein 15. This sequence is 92% identical in mouse and rat.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service