Skip to Content
Merck
All Photos(2)

Documents

K0890

Sigma-Aldrich

Anti-Potassium Channel Kv11.3 (erg3) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KCNH7, Anti-ether-a-go-go-related channel 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable using rat brain sections
western blot: 1:300 using rat brain lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNH7(90134)
mouse ... Kcnh7(170738)
rat ... Kcnh7(170739)

General description

The gene Kcnh7 (potassium voltage-gated channel subfamily H member 7), also referred to as erg3, encodes a subunit of ether-à-go-go related (erg) K+ channels. The erg K+ channels are part of the EAG (ether-à-go-go) family of voltage-gated K+ channels. The encoded protein is found to be expressed in the apical membrane of the retinal pigment epithelium.

Specificity

The KV11.3 antibody is directed against an intracellular epitope of C-terminal part of KV11.3.

Immunogen

synthetic peptide CPEFLDLEKSKLKSKE corresponding to amino acids 1108-1123 of rat KV11.3.

Biochem/physiol Actions

The gene Kcnh7 (potassium voltage-gated channel subfamily H member 7) encodes a voltage gated K+ channel that modulates the membrane potential in its rested and activated state. In the nervous system, it may participate in the regulation of neuronal excitability. The erg3 channels have relatively negative activation threshold compared to erg1 and erg2. They are capable of recycling K+ ions to the subretinal space in the dark and contribute to the maintenance of the resting membrane potential.

Physical form

Lyophilized powder containing phosphate buffered saline, pH 7.4, 1% BSA and 0.025% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional properties of human neuronal Kv11 channels.
Einarsen K
Pflugers Archiv : European Journal of Physiology, 458, 689-700 (2009)
Sönke Cordeiro et al.
PloS one, 6(12), e29490-e29490 (2011-12-30)
In response to light, most retinal neurons exhibit gradual changes in membrane potential. Therefore K+ channels that mediate threshold currents are well-suited for the fine-tuning of signal transduction. In the present study we demonstrate the expression of the different Kv11

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service