Skip to Content
Merck
All Photos(1)

Documents

ILGA2F2S26

Sigma-Aldrich

SILuCarb Glycan Standard 13C Biantennary A2F 2xSial(Alpha-2,6)

13C  labeled di-sialylated, galactosylated biantennary, core-substituted with fucose, Powered by Asparia Glycomics

Synonym(s):

SILuCarb Glycan Standard 13C Biantennary Ab

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8213C8H148N6O66
Molecular Weight:
2378.08
UNSPSC Code:
41105331

shipped in

ambient

storage temp.

2-8°C

General description

SILuCarb Glycan Standards are patented synthetic 13C-labeled N-glycans for use as internal standards in absolute glycan quantification by mass spectrometry.

Application

For use as a quantitative standard in applications such as Matrix-assisted Laser Desorption Ionization-time-of-flight (MALDI-TOF), Electrospray Ionization (ESI) and Liquid Chromatography Mass Spectrometry (LC-MS).

Physical form

13C labeled N-glycan refernce standard. 20 microM quantified solution

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service