Skip to Content
Merck

I6005

Sigma-Aldrich

myo-Inositol hexasulfate hexapotassium salt

Synonym(s):

Inositol hexakissulfate, InsS6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
US$598.00

US$598.00


Availability
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

To order products, please contact your local dealer.

Select a Size

Change View
100 μL
US$598.00

About This Item

Linear Formula:
C6H6O24S6K6
CAS Number:
Molecular Weight:
889.08
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

US$598.00


Availability
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

To order products, please contact your local dealer.

storage temp.

−20°C

SMILES string

[K].OS(=O)(=O)OC1C(OS(O)(=O)=O)C(OS(O)(=O)=O)C(OS(O)(=O)=O)C(OS(O)(=O)=O)C1OS(O)(=O)=O

InChI

1S/C6H12O24S6.K.H/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15;;/h1-6H,(H,7,8,9)(H,10,11,12)(H,13,14,15)(H,16,17,18)(H,19,20,21)(H,22,23,24);;

InChI key

JYVHCOUFXHFSLT-UHFFFAOYSA-N

Biochem/physiol Actions

InsS6 has a molecular footprint and negative surface charge that is similar to that of phytic acid (InsP6), but with lower biological activity. It is used as a control in studies of the cellular actions of phytic acid. It is also a competitive inhibitor of Aspergillus phytase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Oshima
Thrombosis research, 58(3), 243-250 (1990-05-01)
Thrombin-clotting time of fibrinogen was delayed by inositol hexasulfate (IHS). Clotting time of the recalcified plasma was also delayed by IHS. Moreover, when IHS was intravenously injected into anesthetized male rabbits (0.11 mmoles/kg), it delayed clotting time of the recalcified
R Kojima et al.
Japanese journal of pharmacology, 53(3), 347-358 (1990-07-01)
We examined the protective effect of inositol hexasulfate (IS6) against tobramycin (TOB)-induced nephrotoxicity. In the electrophoretic analysis, TOB alone and IS6 alone were observed as single spots on the cathode and anode sides, respectively. However, in the mixture of TOB
N M Dean et al.
The Biochemical journal, 250(2), 493-500 (1988-03-01)
Previous studies demonstrated a multiplicity of isomers of inositol phosphates in GH3 rat pituitary tumour cells. In order to determine their origin, we have investigated the metabolism of radiolabelled inositol phosphates (IPn) in GH3 cell homogenates by using h.p.l.c. I(1,4,5)P3
T Otake et al.
Kansenshogaku zasshi. The Journal of the Japanese Association for Infectious Diseases, 63(7), 676-683 (1989-07-01)
The monosaccharide substances inositol hexasulfate (IHS) and inositol hexaphosphoric acid (Phytic acid, IHP) were investigated for their antiviral effect on the human immunodeficiency virus (HIV) in vitro. In MT-4 cells IHS completely inhibited the cytopathic effect of HIV and the
Aaron J Oakley
Biochemical and biophysical research communications, 397(4), 745-749 (2010-06-15)
Phytases hydrolyse the phosphomonoesters of phytate (myo-inositol-1,2,3,4,5,6-hexakis phosphate) and thus find uses in plant and animal production through the mobilisation of phosphorus from this source. The structure of partially deglycosylated Aspergillus niger PhyA is presented in apo form and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service