I1006
1,2-O-Isopropylidene-α-D-glucofuranose
≥98% (TLC)
Synonym(s):
Monoacetone glucose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H16O6
CAS Number:
Molecular Weight:
220.22
Beilstein:
82670
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
Assay
≥98% (TLC)
storage temp.
−20°C
SMILES string
CC1(C)O[C@H]2O[C@H]([C@H](O)CO)[C@H](O)[C@H]2O1
InChI
1S/C9H16O6/c1-9(2)14-7-5(12)6(4(11)3-10)13-8(7)15-9/h4-8,10-12H,3H2,1-2H3/t4-,5+,6-,7-,8-/m1/s1
InChI key
BGGCXQKYCBBHAH-OZRXBMAMSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Eunsook S Jin et al.
Analytical biochemistry, 327(2), 149-155 (2004-03-31)
A triple-tracer method was developed to provide absolute fluxes contributing to endogenous glucose production and hepatic tricarboxylic acid (TCA) cycle fluxes in 24-h-fasted rats by (2)H and (13)C nuclear magnetic resonance (NMR) analysis of a single glucose derivative. A primed
Eunsook S Jin et al.
American journal of physiology. Endocrinology and metabolism, 288(4), E654-E662 (2004-11-25)
The metabolic mechanism of hepatic glucose overproduction was investigated in 3,3'-5-triiodo-l-thyronine (T3)-treated rats and Zucker diabetic fatty (ZDF) rats (fa/fa) after a 24-h fast. 2H2O and [U-13C3]propionate were administered intraperitoneally, and [3,4-13C2]glucose was administered as a primed infusion for 90
Takashi Kobayashi et al.
Biotechnology letters, 32(11), 1679-1684 (2010-06-25)
In order to synthesize a sugar ester at high concentration, 1,2-O-isopropylidene-α-D-glucofuranose (IpGlc), which is one of the sugar acetals and is more hydrophobic than unmodified glucose, was esterified with palmitic acid at 40°C using immobilized lipase from Candida antarctica in
Fermentation of 1,2-O-iso-propylidene-D-glucofuranose ("monoacetone glucose") by anaerobic bacteria.
S H Cmelik
Zentralblatt fur Bakteriologie. 1. Abt. Originale. A: Medizinische Mikrobiologie, Infektionskrankheiten und Parasitologie, 247(4), 495-501 (1980-01-01)
Various species of Clostridium, Bacteroides, Propionibacterium and Eubacterium were incubated in a 1% solution of 1,2-O-iso-propylidene-D-glucofuranose in a peptone-yeast-extract (PY) medium according to the VPI-technique. The volatile and non volatile acids were investigated by gas-liquid chromatography. All microorganisms showed a
Aifric O'Sullivan et al.
Molecular nutrition & food research, 55(7), 1018-1025 (2011-04-27)
Research reports suggest that vitamin D affects glucose and insulin metabolism; however, the exact mechanisms are unclear. ²H NMR analysis of monoacetone glucose (MAG) after tracer administration provides a non-invasive method of profiling hepatic glucose metabolism. This study examined the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service