Documents
HS86655
Cell Spreader
silver stainless steel, bar L 33 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
silver stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS86655
bar L
33 mm
General description
The Stainless Steel Cell Spreader promotes smooth, controlled spreading.
Features and Benefits
  • Reusable
  • Sterilizable
  • Flame and autoclavable