Skip to Content
Merck
All Photos(2)

Documents

HPA023187

Sigma-Aldrich

Anti-TMEM70 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ20533, Anti-transmembrane protein 70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KGYVIRLYHEATTDTYKAITYNAMLAETSTVFHQNDVKIPDAKHVFTTFYAKTKSLLVNPVLFPNREDYIHLMGYDKEEFILYMEETSEEKRHKDDK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMEM70(54968)

General description

TMEM70 (transmembrane protein 70) is a mitochondrial protein and is present in the inner mitochondrial membrane. It is generated as a 29kDa precursor and is processed to a 21kDa protein. The gene is mapped to human chromosome 8q21.

Immunogen

transmembrane protein 70 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TMEM70 (transmembrane protein 70) is a helper protein of the mitochondrial complex V (F1F0-ATP synthase). Mutations in TMEM70 result in nuclear ATP synthase deficiency, which is associated with neonatal hypotonia, hypertrophic cardiomyopathy, facial dysmorphism, lactic acidosis and 3-methylglutaconic aciduria.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72939

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michela Catteruccia et al.
Molecular genetics and metabolism, 111(3), 353-359 (2014-02-04)
Mutations in the TMEM70 are the most common cause of nuclear ATP synthase deficiency resulting in a distinctive phenotype characterized by severe neonatal hypotonia, hypertrophic cardiomyopathy (HCMP), facial dysmorphism, severe lactic acidosis, hyperammonemia and 3-methylglutaconic aciduria (3-MGA). We collected 9
Matthias Choschzick et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 23(4), 603-610 (2010-02-09)
Copy number gains involving the long arm of chromosome 8, including high-level amplifications at 8q21 and 8q24, have been frequently reported in breast cancer. Although the role of the MYC gene as the driver of the 8q24 amplicon is well
Vendula Havlíčková Karbanová et al.
Biochimica et biophysica acta, 1817(7), 1037-1043 (2012-03-22)
Early onset mitochondrial encephalo-cardiomyopathy due to isolated deficiency of ATP synthase is frequently caused by mutations in TMEM70 gene encoding enzyme-specific ancillary factor. Diminished ATP synthase results in low ATP production, elevated mitochondrial membrane potential and increased ROS production. To
Alessandra Torraco et al.
Neurogenetics, 13(4), 375-386 (2012-09-19)
Mammalian complex V (F1F0-ATP synthase or ATPase) uses the proton gradient to generate ATP during oxidative phosphorylation and requires several helper proteins, including TMEM70, to form the holoenzyme in a stepwise process in which nuclear DNA is combined with mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service