Skip to Content
Merck
All Photos(5)

Key Documents

HPA004694

Sigma-Aldrich

Anti-ARSD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASD antibody produced in rabbit, Anti-Arylsulfatase D precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

QGAEARSAHEFLFHYCGQHLHAARWHQKDSGSVWKVHYTTPQFHPEGAGACYGRGVCPCSGEGVTHHRPPLLFDLSRDPSEARPLTPDSEPLYHAVIARVGAAVSEHRQTLSPVPQQFSMSNILWKPWLQPCCGHFPFCSCHED

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARSD(414)

General description

ARSD (arylsulfatase D) belongs to the sulfatase gene family. It is a homodimer composed of three adjacent cysteine pairs i.e. Cys12-Cys13, Cys112-Cys113 and Cys119-Cys120 in each subunit. It acts as an arsenic chaperone in the ArsAB (arsenite-antimonite) pump system. Its cysteine pair binds to the As(III) or Sb(III) for transferring them to the ArsA subunit of the pump. ARSD is involved in the sphingolipid metabolism.

Immunogen

Arylsulfatase D precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ARSD antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74273

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a diagnostic algorithm in periodontal disease and identification of genetic expression patterns: A preliminary report.
Garzon I
Journal of Dentistry, 7(1), 48-56 (2012)
Yung-Feng Lin et al.
Journal of bioenergetics and biomembranes, 39(5-6), 453-458 (2007-10-24)
The toxic metalloid arsenic is widely disseminated in the environment and causes a variety of health and environment problems. As an adaptation to arsenic-contaminated environments, organisms have developed resistance systems. Many ars operons contain only three genes, arsRBC. Five gene
I Garzón et al.
Journal of tissue engineering and regenerative medicine, 12(1), 204-216 (2017-01-13)
Regeneration of the pulp-dentin complex with stem cells is a potential alternative to conventional root canal treatments. Human dental pulp stem cells (hDPSCs) have been extensively studied because of their ability to proliferate and differentiate into mineralized dental and non-dental
G Meroni et al.
Human molecular genetics, 5(4), 423-431 (1996-04-01)
An obligatory crossing-over event between the X and Y chromosomes in mammals occurs at each male meiosis within the 2.6 Mb of DNA defining the pseudoautosomal region (PAR). Genes located within or near the human PAR have homologous copies on
Alessandra Trojani et al.
Cancer biomarkers : section A of Disease markers, 11(1), 15-28 (2011-01-01)
Several studies demonstrated IGVH mutational status and ZAP70 expression as the most relevant prognostic markers in Chronic Lymphocytic Leukemia (CLL), suggesting the separation of two patient subgroups: with good mutated ZAP70 negative (MTZAP70(-) and poor unmutated ZAP70 positive (UMZAP70(+)) prognosis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service