Skip to Content
Merck
All Photos(2)

Key Documents

H6271

Sigma-Aldrich

(±)-2-Hydroxydecanoic acid

≥98% (capillary GC)

Synonym(s):

α-Hydroxycapric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O3
CAS Number:
Molecular Weight:
188.26
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98% (capillary GC)

storage temp.

2-8°C

SMILES string

CCCCCCCCC(O)C(O)=O

InChI

1S/C10H20O3/c1-2-3-4-5-6-7-8-9(11)10(12)13/h9,11H,2-8H2,1H3,(H,12,13)

InChI key

GHPVDCPCKSNJDR-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Takahashi et al.
Pharmaceutical research, 11(3), 388-392 (1994-03-01)
The enhancing action of decanoic acid (C10) and its derivatives on mucosal absorption of phenolsulfonphthalein (PSP) in the jejunum or colon was analyzed using pharmacokinetics in rats. After administration of a solution containing PSP and an enhancer [C10, 2-hydroxydecanoic acid
Gerald Nagahashi et al.
Fungal biology, 115(4-5), 351-358 (2011-05-03)
Two hydroxy fatty acids, tentatively identified previously in carrot root exudates, were tested for their effects on hyphal growth of the arbuscular mycorrhizal (AM) fungus, Gigaspora gigantea (Nicol. and Gerd.) Gerdemann and Trappe. Best results were achieved with a long-term bioassay
Liya Ge et al.
Talanta, 146, 107-113 (2015-12-24)
Polyhydroxyalkanoates (PHAs) are commercially-valuable biocompatible and biodegradable polymers with many potential medical, pharmaceutical and other industrial applications. The analysis of PHA monomeric composition is especially challenging due to the broad chemical diversity of PHA monomers and lack of analytical standards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service